Learn More
NanoScreen LLC TIPS 30UL 384 PE COMP 19200CS
Catalog No. 50921310
Encompass_Preferred
Pipette tips, Robotic. PerkinElmer P30, 384 tips. Sterile Tips. For PerkinElmer/Packard 384 dispense heads found on MiniTrak, Janus and EP3. Tips are 100% inspected to ensure quality. Guaranteed 1536 compatible. Case=50 racks=19200 tips
Catalog No. 50-921-310 Supplier NanoScreen LLC Supplier No. TIPP30384SA
Pipette tips, Robotic. PerkinElmer P30, 384 tips. Sterile Tips. For PerkinElmer/Packard 384 dispense heads found on MiniTrak, Janus and EP3. Tips are 100% inspected to ensure quality. Guaranteed 1536 compatible. Case=50 racks=19200 tips