Learn More
NanoScreen LLC TIPS 30UL SQ 96 S PE COMPATIBL
Catalog No. 50921306
Encompass_Preferred
Pipette tips, Robotic. PerkinElmer/CCS P30, 96 tips. Sterile Tips. Designed to work with PerkinElmer/Packard 384 dispense heads found on MiniTrak, Janus and EP3. Tips are racked in a single quadrant, 96 tips/rack Case=50 racks=4800 tips
Catalog No. 50-921-306 Supplier NanoScreen LLC Supplier No. TIPP30384SQ
Pipette tips, Robotic. PerkinElmer/CCS P30, 96 tips. Sterile Tips. Designed to work with PerkinElmer/Packard 384 dispense heads found on MiniTrak, Janus and EP3. Tips are racked in a single quadrant, 96 tips/rack Case=50 racks=4800 tips