Learn More
Beckman Coulter Beckman Coulter Biomek i-Series 50 μL Tips, Non-sterile - 3,840 tips (10 racks of 384) - B85753
Catalog No. NC2752086 Shop All Beckman Coulter Products
Encompass_Preferred
Beckman Coulter Biomek i-Series 50 μL non-sterile tips (MPN B85753) are engineered for automated liquid-handling on Biomek i-Series workstations. Supplied in rack format for high-throughput workflows, they provide consistent volume delivery for routine pipetting applications where sterile tips are not required.
  • Package quantity: 10 racks per case.
  • Tips per case: 3,840 tips.
  • Tips per rack: 384 per rack.
  • Tip volume: 50 μL.
  • Sterility: Non-sterile.
  • Rack format: 384 tips per rack.
  • BioSafe bio-certified.
  • Nonpyrogenic.
Catalog No. NC2752086 Supplier Beckman Coulter Supplier No. B85753
Beckman Coulter Biomek i-Series 50 μL non-sterile tips (MPN B85753) are engineered for automated liquid-handling on Biomek i-Series workstations. Supplied in rack format for high-throughput workflows, they provide consistent volume delivery for routine pipetting applications where sterile tips are not required.
  • Package quantity: 10 racks per case.
  • Tips per case: 3,840 tips.
  • Tips per rack: 384 per rack.
  • Tip volume: 50 μL.
  • Sterility: Non-sterile.
  • Rack format: 384 tips per rack.
  • BioSafe bio-certified.
  • Nonpyrogenic.