Learn More
Andwin Scientific Universal Pipet Tips, 20-200 uL, Nonsterile, 960/pk
SDP
Catalog No. NC1809941
Encompass_Preferred
Universal low retention pipet tips repel liquid by creating a hydrophobic surface on the inside of the tip increasing your accuracy and data quality.
Catalog No. NC1809941 Supplier Andwin Scientific Supplier No. 76322144
Universal low retention pipet tips repel liquid by creating a hydrophobic surface on the inside of the tip increasing your accuracy and data quality.