Learn More
Med Vet International Dynarex Pre Bent Flow Tips, 22g - Sealant-Cement Tips, Gray, 2500/Case
Catalog No. 502831878
Encompass_Preferred
The Dynarex Pre-Bent Flow Tips are optimal for precise placement and dispensing flow for any dental procedure. These Dental Flow Tips are available in 4 different tips and gauges appropriate to different material types and are color-coded for easy distinction. The Dynarex Dental Flow Tips are perfect for use with resins, etchants, and other flowable composites. Dental Pre-Bent Flow Tips are designed with various applications in mind, with four gauges allowing you to dispense flow sealant, etch, liner and sealant cement.
Catalog No. 50-283-1878 Supplier Med Vet International Supplier No. 2214
The Dynarex Pre-Bent Flow Tips are optimal for precise placement and dispensing flow for any dental procedure. These Dental Flow Tips are available in 4 different tips and gauges appropriate to different material types and are color-coded for easy distinction. The Dynarex Dental Flow Tips are perfect for use with resins, etchants, and other flowable composites. Dental Pre-Bent Flow Tips are designed with various applications in mind, with four gauges allowing you to dispense flow sealant, etch, liner and sealant cement.