Learn More
Andwin Scientific Universal Fit Tips, Low Retention, 10 uL, 960/pk
SDP
Supplier:  Andwin Scientific 89140160
Encompass_Preferred
Pre-sterile universal barrier tips are made from virgin polypropylene and feature high density polyethylene filters to ensure against aerosol contamination and provide superior consistency.
Catalog No. 50-112-6041