Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Tips, Nonsterile

Catalog No. 01677202 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
1000 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01-677-202 10 μL
01-677-200 200 μL
01-677-201 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-677-202 Supplier Corning™ Supplier No. P37106072
Add to Cart
Add to Cart

Easy to open hinged lids with a sturdy closing mechanism to keep tips free of contamination.

  • Empty pipette tip racks designed for pipette tip reloads filter pipette tip reloads and bulk pipette tips
  • Easy to open hinged lids with a sturdy closing mechanism to help keep tips free of contamination
  • Available for 10 μL 200 μL and 1000 μL tips
  • RNaseDNase free
  • Human DNA free
  • NonPyrogenic
  • ATP free
  • Manufactured in a facility that complies with the current revision of ISO 9001 standard
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Gray
Autoclavable Not Autoclavable
Format Reload Rack (tips not included)
Tip Style Standard
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.