Learn More
GREENLABS LLC DBA GREENLABS RECYCLING Tips Transfer Bin manufactured in Massachusetts from 100% recycled lab plastic
Catalog No. NC2939826
Encompass
This .95 gallon recycled tips transfer bin is designed for reusability on the bench. The sliding lid door can be secured to create a closed system and re-opened. The opening can accommodate a multi-channel pipette. The lid itself can be removed from the bin’s base. The design enables you to remove the lid and transfer materials into a larger hazardous waste receptacle and reuse the bin. This product was designed and manufactured in Massachusetts made of recycled material collected from Boston area laboratories. Each product will be uniquely colored and no dyes are added.
Catalog No. NC2939826 Supplier GREENLABS LLC DBA GREENLABS RECYCLING Supplier No. 001
This .95 gallon recycled tips transfer bin is designed for reusability on the bench. The sliding lid door can be secured to create a closed system and re-opened. The opening can accommodate a multi-channel pipette. The lid itself can be removed from the bin’s base. The design enables you to remove the lid and transfer materials into a larger hazardous waste receptacle and reuse the bin. This product was designed and manufactured in Massachusetts made of recycled material collected from Boston area laboratories. Each product will be uniquely colored and no dyes are added.