Learn More
Tecan 50L MCA96 Tip NFil
Catalog No. NC0248608
Encompass_Preferred
Genuine Tecan 50 L disposable automation tips designed specifically for use on the Freedom EVO and Fluent MultiChannel Arm™ (MCA) 96 option. ANSI format, single stack, non-filtered. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. 40 tip boxes x 96 tips, 3,840 tips/sales unit. All MultiChannel Arm™(MCA) 96 disposable tips are designed, manufactured, tested and certified to guarantee reliable quality, helping you to acquire optimal results.
This item is not returnable. View return policy
Catalog No. NC0248608 Supplier Tecan Supplier No. 30038606
Genuine Tecan 50 L disposable automation tips designed specifically for use on the Freedom EVO and Fluent MultiChannel Arm™ (MCA) 96 option. ANSI format, single stack, non-filtered. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. 40 tip boxes x 96 tips, 3,840 tips/sales unit. All MultiChannel Arm™(MCA) 96 disposable tips are designed, manufactured, tested and certified to guarantee reliable quality, helping you to acquire optimal results.