Learn More
Andwin Scientific TIPSTTN 1-200UL NTLWST 5PK/CS
SDP
Catalog No. NC0278694
Encompass_Preferred
The TipStation Pipet Tip Systems are compatible with 8- and 12-channel pipettors. The racks have a stackable design that allow users to save bench space and utilize it for other purposes. Racks can also be snapped together and not lift during the process of pipetting. The wells are recessed that help in providing stability for mouting on pipettors.
Catalog No. NC0278694 Supplier Andwin Scientific Supplier No. 27770T
The TipStation Pipet Tip Systems are compatible with 8- and 12-channel pipettors. The racks have a stackable design that allow users to save bench space and utilize it for other purposes. Racks can also be snapped together and not lift during the process of pipetting. The wells are recessed that help in providing stability for mouting on pipettors.