Learn More
Enzo Life Sciences TIQ-A (5mg). CAS: 420849-22-5
SDP
Supplier:  Enzo Life Sciences ALX270365M005
Encompass_Preferred
Potent poly(ADP-ribose) polymerase-1 (PARP-1) inhibitor (IC50=450nM). Neuroprotectant. Alternative name: Thieno[2,3-c]isoquinolin-5-one. Formula: C11H7NOS. MW: 201.2. Purity: ≥96% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: +4°C.
Catalog No. 50-200-8511