Learn More
Azzota Corp TIR-05 Ceramic Stirrer 5000ml
Catalog No. 5000887299
Encompass_Preferred
TIR-05 Ceramic Magnetic Stirrer up to 5000ml 200-2000rpm Max 662F(380C) 190x190mm hot plate 110V 600W
Catalog No. 50-008-87299 Supplier Azzota Corp Supplier No. TIR05
TIR-05 Ceramic Magnetic Stirrer up to 5000ml 200-2000rpm Max 662F(380C) 190x190mm hot plate 110V 600W