Learn More
Azzota Corp TIR-5D Digital Stirrer 5000ml
Catalog No. 5000887265
Encompass_Preferred
TIR-5D Digital Ceramic Magnetic Stirrer up to 5000ml 200-2000rpm Max 662F(350C) 190x190mm hot plate 110V 600W
Catalog No. 50-008-87265 Supplier Azzota Corp Supplier No. TIR5D
TIR-5D Digital Ceramic Magnetic Stirrer up to 5000ml 200-2000rpm Max 662F(350C) 190x190mm hot plate 110V 600W