Learn More
Selleck Chemical LLC tirabrutinib
SDP
Catalog No. 5000000045
Encompass_Preferred
Tirabrutinib Hydrochloride (ONO-4059 GS-4059) is a highly potent and selective BTK inhibitor with an IC50 of 2 2 nM
Catalog No. 50-000-00045 Supplier Selleck Chemical LLC Supplier No. S81665MG
Tirabrutinib Hydrochloride (ONO-4059 GS-4059) is a highly potent and selective BTK inhibitor with an IC50 of 2 2 nM