Learn More
TARGETMOL CHEMICALS INC TIRABRUTINIB 5MG
SDP
Catalog No. 502578810
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tirabrutinib (GS-4059) (ONO-4059) is an orally active Bruton’s Tyrosine Kinase (Btk) inhibitor (can cross the blood-brain barrier (BBB)) with an IC50 of 6.8 nM. Tirabrutinib irreversibly and covalently binds to Btk and inhibits aberrant B cell receptor signaling. Tirabrutinib can be used in studies of autoimmune diseases and hematological malignancies. purity: 99%
Catalog No. 50-257-8810 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ02365MG
Add to Cart
Add to Cart
Also available in 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tirabrutinib (GS-4059) (ONO-4059) is an orally active Bruton’s Tyrosine Kinase (Btk) inhibitor (can cross the blood-brain barrier (BBB)) with an IC50 of 6.8 nM. Tirabrutinib irreversibly and covalently binds to Btk and inhibits aberrant B cell receptor signaling. Tirabrutinib can be used in studies of autoimmune diseases and hematological malignancies. purity: 99%