Learn More
Medchemexpress LLC HY-15771A 5mg Medchemexpress, Tirabrutinib (hydrochloride) CAS:1439901-97-9 Purity:>98%
SDP
Catalog No. 502030679
Encompass_Preferred
Medchemexpress, HY-15771A 5mg Tirabrutinib (hydrochloride) CAS:1439901-97-9 Tirabrutinib (ONO-4059) hydrochloride is a selective and novel inhibitor of BTK with IC50 2.2 nm, Tirabrutinib binds to BTK within B cells, thereby preventing B-cell receptor signaling and impeding B-cell development. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0679 Supplier Medchemexpress LLC Supplier No. HY15771A5MG
Medchemexpress, HY-15771A 5mg Tirabrutinib (hydrochloride) CAS:1439901-97-9 Tirabrutinib (ONO-4059) hydrochloride is a selective and novel inhibitor of BTK with IC50 2.2 nm, Tirabrutinib binds to BTK within B cells, thereby preventing B-cell receptor signaling and impeding B-cell development. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.