Learn More
Aviva Systems Biology Corp TIRAP Antibody - middle region (ARP52870_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52870_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TIRAP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TIRAP. Alternate names: FLJ42305, Mal, wyatt, BACTS1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60857