Learn More
Cayman Chemical Tirapazamine, Purity: ≥98%, CAS: 27314-97-2, 100 mg
SDP
Catalog No. 502936326
Encompass_Preferred
A hypoxia-activated anticancer agent; is converted to an oxidizing radical under hypoxic conditions; can induce single- and double-strand breaks in DNA, formation of trapped topoisomerase I- and II-DNA complexes, and other chromosomal aberrations, leading to cytotoxicity; induces hypoxia-enhanced cytotoxicity in DT40 cells (IC50s = 1.02 and 4.34 μM in hypoxic and normoxic conditions, respectively); induces cytotoxicity in HT-1080 and A549 cells in a concentration-dependent manner under hypoxic conditions; sensitizes tumors to cisplatin in a mouse RIF-1 fibrosarcoma tumor model; sensitizes tumors to fractionated irradiation in a FaDu hypopharyngeal squamous cell carcinoma mouse xenograft model at (0.08 mmol/kg)
Catalog No. 50-293-6326 Supplier Cayman Chemical Supplier No. 25981100 MG
A hypoxia-activated anticancer agent; is converted to an oxidizing radical under hypoxic conditions; can induce single- and double-strand breaks in DNA, formation of trapped topoisomerase I- and II-DNA complexes, and other chromosomal aberrations, leading to cytotoxicity; induces hypoxia-enhanced cytotoxicity in DT40 cells (IC50s = 1.02 and 4.34 μM in hypoxic and normoxic conditions, respectively); induces cytotoxicity in HT-1080 and A549 cells in a concentration-dependent manner under hypoxic conditions; sensitizes tumors to cisplatin in a mouse RIF-1 fibrosarcoma tumor model; sensitizes tumors to fractionated irradiation in a FaDu hypopharyngeal squamous cell carcinoma mouse xenograft model at (0.08 mmol/kg)