Learn More
Apexbio Technology LLC Tirapazamine(Synonyms: SR-4233, WIN 59075, Tirazone, TPZ, 3-Amino-1,2,4-benzotriazine 1,4-dioxide), 10mM (in 1mL DMSO), CAS: 27314-97-2.
SDP
Catalog No. 5000060217
Encompass_Preferred
Tirapazamine (CAS 27314-97-2) also known as SR259075 Win59075 or SR4233 is an investigational anticancer agent selectively activated under hypoxic conditions characteristic of solid tumors The compound is bioreduced in hypoxic tumor microenvironments to cytotoxic free radicals leading to cellular damage Tirapazamine downregulates hypoxia-inducible factor-1 (HIF-1 ) thereby influencing pathways associated with tumor adaptive mechanisms to hypoxia Combined treatment with tirapazamine and topoisomerase I inhibitors (such as SN-38 the active metabolite of irinotecan) enhances apoptosis via mitochondria-mediated caspase activation and weakened DNA repair signaling Preclinical investigations including hepatocellular carcinoma xenografts confirm this enhanced tumor suppression Clinical trials evaluating tirapazamine s anticancer efficacy are ongoing
Catalog No. 50-000-60217 Supplier Apexbio Technology LLC Supplier No. B33995.1
Tirapazamine (CAS 27314-97-2) also known as SR259075 Win59075 or SR4233 is an investigational anticancer agent selectively activated under hypoxic conditions characteristic of solid tumors The compound is bioreduced in hypoxic tumor microenvironments to cytotoxic free radicals leading to cellular damage Tirapazamine downregulates hypoxia-inducible factor-1 (HIF-1 ) thereby influencing pathways associated with tumor adaptive mechanisms to hypoxia Combined treatment with tirapazamine and topoisomerase I inhibitors (such as SN-38 the active metabolite of irinotecan) enhances apoptosis via mitochondria-mediated caspase activation and weakened DNA repair signaling Preclinical investigations including hepatocellular carcinoma xenografts confirm this enhanced tumor suppression Clinical trials evaluating tirapazamine s anticancer efficacy are ongoing