Learn More
Medchemexpress LLC Tirapazamine | 27314-97-2 | 99.0% | 178.15 | 200 MG
SDP
Catalog No. 5000351437
Encompass_Preferred
Tirapazamine (SR259075) is an anticancer agent that shows selective cytotoxicity for hypoxic cells in solid tumors, thereby inducing single- and double-strand breaks in DNA, base damage, and cell death. It is an anticancer and bioreductive agent that can enhance the cytotoxic effects of ionizing radiation in hypoxic cells.
  • Selective cytotoxicity for hypoxic cells
  • Induces single- and double-strand breaks in DNA
  • Induces base damage and cell death
  • Enhances cytotoxic effects of ionizing radiation in hypoxic cells
Catalog No. 50-003-51437 Supplier Medchemexpress LLC Supplier No. HY13767200MG
Tirapazamine (SR259075) is an anticancer agent that shows selective cytotoxicity for hypoxic cells in solid tumors, thereby inducing single- and double-strand breaks in DNA, base damage, and cell death. It is an anticancer and bioreductive agent that can enhance the cytotoxic effects of ionizing radiation in hypoxic cells.
  • Selective cytotoxicity for hypoxic cells
  • Induces single- and double-strand breaks in DNA
  • Induces base damage and cell death
  • Enhances cytotoxic effects of ionizing radiation in hypoxic cells