Learn More
TARGETMOL CHEMICALS INC TIRBANIBULIN 25MG
SDP
Catalog No. 502579421
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tirbanibulin (KX2-391) is a highly selective Src kinase inhibitor that has demonstrated efficacy in pre-Clinicalal animal models of colon pancreatic prostate and breast cancer. It is a substrate-targeted kinase inhibitor. KX2-391 belongs to an emerging new family of targeted cancer treatments called protein kinase inhibitors. purity: 99%
Catalog No. 50-257-9421 Supplier TARGETMOL CHEMICALS INC Supplier No. T634525MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tirbanibulin (KX2-391) is a highly selective Src kinase inhibitor that has demonstrated efficacy in pre-Clinicalal animal models of colon pancreatic prostate and breast cancer. It is a substrate-targeted kinase inhibitor. KX2-391 belongs to an emerging new family of targeted cancer treatments called protein kinase inhibitors. purity: 99%