Learn More
Medchemexpress LLC HY-10340 5mg Medchemexpress, Tirbanibulin CAS:897016-82-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103405MG
Encompass_Preferred
Medchemexpress, HY-10340 5mg Tirbanibulin CAS:897016-82-9 Tirbanibulin (KX2-391) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0670