Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tirbanibulin dihydrochloride | 1038395-65-1 | MFCD18633218 | 98.0% | 504.45 | C26H31Cl2N3O3 | 10 MG
SDP

Catalog No. 5000244908
Encompass_Preferred

Tirbanibulin dihydrochloride is the dihydrochloride salt of tirbanibulin, a peptidomimetic Src inhibitor that targets the peptide-substrate site of Src and demonstrates GI50 values of approximately 9-60 nM in cancer cell lines. This product is supplied as a purified analytical standard for research and analytical applications.

  • Src inhibitor targeting the peptide-substrate site
  • Dihydrochloride salt form for improved handling and solubility
  • Supplied as a solid analytical standard for research use
  • Reported purity ~98.0%
  • Molecular weight 504.45; formula C26H31Cl2N3O3
  • CAS number 1038395-65-1
  • Suitable for in vitro and analytical assays

Catalog No. 50-002-44908 Supplier Medchemexpress LLC Supplier No. HY10340A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tirbanibulin dihydrochloride is the dihydrochloride salt of tirbanibulin, a peptidomimetic Src inhibitor that targets the peptide-substrate site of Src and demonstrates GI50 values of approximately 9-60 nM in cancer cell lines. This product is supplied as a purified analytical standard for research and analytical applications.

  • Src inhibitor targeting the peptide-substrate site
  • Dihydrochloride salt form for improved handling and solubility
  • Supplied as a solid analytical standard for research use
  • Reported purity ~98.0%
  • Molecular weight 504.45; formula C26H31Cl2N3O3
  • CAS number 1038395-65-1
  • Suitable for in vitro and analytical assays

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.