Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tirbanibulin dihydrochloride | 1038395-65-1 | MFCD20526429 | 98.0% | 504.45 g/mol | C26H31Cl2N3O3 | 50 MG
SDP

Catalog No. 5000245130
Encompass_Preferred

Tirbanibulin dihydrochloride is a research-grade small-molecule inhibitor that targets Src kinase and microtubule/tubulin dynamics. Supplied as a solid for in vitro and preclinical laboratory studies, it is provided with analytical characterization and storage guidance. For research use only; not for human therapeutic use.

  • Targets Src and microtubule/tubulin.
  • Cas number: 1038395-65-1.
  • Molecular formula: C26H31Cl2N3O3.
  • Molecular weight: 504.45 g/mol.
  • Purity: 98.02% (reported).
  • Appearance: solid; recommended storage 4°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Supplied as a solid; common package size: 50 MG.

Catalog No. 50-002-45130 Supplier Medchemexpress LLC Supplier No. HY10340A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tirbanibulin dihydrochloride is a research-grade small-molecule inhibitor that targets Src kinase and microtubule/tubulin dynamics. Supplied as a solid for in vitro and preclinical laboratory studies, it is provided with analytical characterization and storage guidance. For research use only; not for human therapeutic use.

  • Targets Src and microtubule/tubulin.
  • Cas number: 1038395-65-1.
  • Molecular formula: C26H31Cl2N3O3.
  • Molecular weight: 504.45 g/mol.
  • Purity: 98.02% (reported).
  • Appearance: solid; recommended storage 4°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Supplied as a solid; common package size: 50 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.