Learn More
TARGETMOL CHEMICALS INC Tirbanibulin Mesylate 25MG
SDP
Catalog No. 502876748
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Tirbanibulin Mesylate (KX01 Mesylate) is an inhibitor of Src that targets the peptide substrate site of Src (GI50 9-60 nM in cancer cell lines). Purity 99.42%
Catalog No. 50-287-6748 Supplier TARGETMOL CHEMICALS INC Supplier No. T1567525MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Tirbanibulin Mesylate (KX01 Mesylate) is an inhibitor of Src that targets the peptide substrate site of Src (GI50 9-60 nM in cancer cell lines). Purity 99.42%