Learn More
Medchemexpress LLC HY-10340B 5mg Medchemexpress, Tirbanibulin (Mesylate) CAS:1080645-95-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10340B5MG
Encompass_Preferred
Medchemexpress, HY-10340B 5mg Tirbanibulin (Mesylate) CAS:1080645-95-9 Tirbanibulin Mesylate (KX2-391 Mesylate) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8102