Learn More
Mopec Tire and Footprint Casting Release Spray
SDP
Catalog No. 22446313 Shop All Mopec Products
Change view
Click to view available options
Quantity:
9 oz.
Catalog No. Quantity
22-446-313 9 oz.
1 options
Catalog No. 22-446-313 Supplier Mopec Supplier No. FS390
Add to Cart
Add to Cart
Specially formulated to prevent casting materials from sticking to molds or surfaces, this spray creates a thin, even release barrier. It allows tire and footprint casts to separate more easily while helping preserve fine impression details and supporting clean, detailed casting results.
Performance
  • Aerosol can
Capacity
  • 9 oz.
Performance
  • Helps in the separation of casting materials from the impression
Specifications
Quantity 9 oz.