Learn More
INTERWORLD HIGHWAY LLC Mitutoyo Absolute Digimatic Depth Gage - Tire Tread, Base 2.36in, Range 0-1in
SDP
Catalog No. NC3660711
Encompass
The Mitutoyo ABSOLUTE Digimatic tire tread depth gage is specially designed to measure tire tread depth with accuracy down to +/- 0.02mm/.0005in. Its size and light weight makes the gage easy to use. The gage scale, blade and measuring rod are constructed of hardened stainless steel for added durability. Measurements are displayed on a digital LCD readout with SPC output for easy automatic recording.
Catalog No. NC3660711 Supplier INTERWORLD HIGHWAY LLC Supplier No. 57110020
The Mitutoyo ABSOLUTE Digimatic tire tread depth gage is specially designed to measure tire tread depth with accuracy down to +/- 0.02mm/.0005in. Its size and light weight makes the gage easy to use. The gage scale, blade and measuring rod are constructed of hardened stainless steel for added durability. Measurements are displayed on a digital LCD readout with SPC output for easy automatic recording.