Learn More
TARGETMOL CHEMICALS INC TIRILAZAD MESYLATE 5MG
SDP
Catalog No. 502577031
Encompass_Preferred
Also available in 50mg 100mg and bulk. Please contact Fisher for quotes. Tirilazad mesylate (U 74006F) is a non-glucocorticoid lipid peroxidation inhibitor with antiviral activity and neuroprotective activity.Tirilazad mesylate confines intracellular intramembranous foci attenuates spinal cord injury induced by trauma stroke and ischemia/reperfusion injuries inhibits toxins in dogs reduces the effects of Tirilazad mesylate is used in the study of neurological disorders. purity: 99%
Catalog No. 50-257-7031 Supplier TARGETMOL CHEMICALS INC Supplier No. T289785MG
Also available in 50mg 100mg and bulk. Please contact Fisher for quotes. Tirilazad mesylate (U 74006F) is a non-glucocorticoid lipid peroxidation inhibitor with antiviral activity and neuroprotective activity.Tirilazad mesylate confines intracellular intramembranous foci attenuates spinal cord injury induced by trauma stroke and ischemia/reperfusion injuries inhibits toxins in dogs reduces the effects of Tirilazad mesylate is used in the study of neurological disorders. purity: 99%