Learn More
AdipoGen Tirofiban
SDP
Catalog No. 502559977
Encompass_Preferred
Chemical. CAS 144494-65-5. Formula C22H36N2O5S. MW 440.6. Synthetic Synthetic non-peptide selective glycoprotein GP IIb/IIIa receptor antagonist/inhibitor. Anticoagulant, specifically, an anti-platelet aggregation inhibitor.
Catalog No. 50-255-9977 Supplier AdipoGen Supplier No. CDXT0153M005
Chemical. CAS 144494-65-5. Formula C22H36N2O5S. MW 440.6. Synthetic Synthetic non-peptide selective glycoprotein GP IIb/IIIa receptor antagonist/inhibitor. Anticoagulant, specifically, an anti-platelet aggregation inhibitor.