Learn More
Medchemexpress LLC Tirofiban | 144494-65-5 | 440.60 | 5 MG
SDP
Catalog No. 5000384644
Encompass_Preferred
Tirofiban is a selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist. It inhibits fibrinogen binding to this receptor, providing antithrombotic activity. Tirofiban induces proliferation and migration on endothelial cells by stimulating VEGF production. It can significantly reduce myocardial no-reflow and ischemia-reperfusion injury by alleviating myocardial microvascular structural and endothelial dysfunction in the ischemic area.
  • Selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist
  • Inhibits fibrinogen binding
  • Provides antithrombotic activity
  • Induces endothelial cell proliferation and migration
  • Stimulates vascular endothelial growth factor (VEGF) production
  • Reduces myocardial no-reflow
  • Alleviates myocardial microvascular structural and endothelial dysfunction
  • Targets integrin
  • Affects cytoskeleton pathway
Catalog No. 50-003-84644 Supplier Medchemexpress LLC Supplier No. HY17369B5MG
Tirofiban is a selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist. It inhibits fibrinogen binding to this receptor, providing antithrombotic activity. Tirofiban induces proliferation and migration on endothelial cells by stimulating VEGF production. It can significantly reduce myocardial no-reflow and ischemia-reperfusion injury by alleviating myocardial microvascular structural and endothelial dysfunction in the ischemic area.
  • Selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist
  • Inhibits fibrinogen binding
  • Provides antithrombotic activity
  • Induces endothelial cell proliferation and migration
  • Stimulates vascular endothelial growth factor (VEGF) production
  • Reduces myocardial no-reflow
  • Alleviates myocardial microvascular structural and endothelial dysfunction
  • Targets integrin
  • Affects cytoskeleton pathway