Learn More
Medchemexpress LLC Tirofiban | 144494-65-5 | 98.4% | 440.60 | 50 MG
SDP
Catalog No. 5000385809
Encompass_Preferred
Tirofiban (L700462) is a selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist. It inhibits fibrinogen binding to this receptor, leading to antithrombotic activity. It also induces proliferation and migration of endothelial cells by stimulating VEGF production, and significantly reduces myocardial no-reflow and ischemia-reperfusion injury by alleviating myocardial microvascular structural and endothelial dysfunction in the ischemic area.
  • Selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist
  • Inhibits fibrinogen binding to the receptor
  • Possesses antithrombotic activity
  • Induces proliferation and migration of endothelial cells
  • Reduces myocardial no-reflow and ischemia-reperfusion injury
  • Alleviates myocardial microvascular structural and endothelial dysfunction
Catalog No. 50-003-85809 Supplier Medchemexpress LLC Supplier No. HY17369B50MG
Tirofiban (L700462) is a selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist. It inhibits fibrinogen binding to this receptor, leading to antithrombotic activity. It also induces proliferation and migration of endothelial cells by stimulating VEGF production, and significantly reduces myocardial no-reflow and ischemia-reperfusion injury by alleviating myocardial microvascular structural and endothelial dysfunction in the ischemic area.
  • Selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist
  • Inhibits fibrinogen binding to the receptor
  • Possesses antithrombotic activity
  • Induces proliferation and migration of endothelial cells
  • Reduces myocardial no-reflow and ischemia-reperfusion injury
  • Alleviates myocardial microvascular structural and endothelial dysfunction