Learn More
Selleck Chemical LLC Tirofiban S3085-25mg
SDP
Catalog No. 503109711
Encompass_Preferred
Tirofiban (MK-383) is a selective platelet GPIIb/IIIa antagonist which inhibits platelet aggregation with IC50 of 9 nM
Catalog No. 50-310-9711 Supplier Selleck Chemical LLC Supplier No. S308525MG
Tirofiban (MK-383) is a selective platelet GPIIb/IIIa antagonist which inhibits platelet aggregation with IC50 of 9 nM