Learn More
Selleck Chemical LLC Tirofiban S3085-5mg
SDP
Catalog No. 503109712
Encompass_Preferred
Tirofiban (MK-383) is a selective platelet GPIIb/IIIa antagonist which inhibits platelet aggregation with IC50 of 9 nM
Catalog No. 50-310-9712 Supplier Selleck Chemical LLC Supplier No. S30855MG
Tirofiban (MK-383) is a selective platelet GPIIb/IIIa antagonist which inhibits platelet aggregation with IC50 of 9 nM