Learn More
TARGETMOL CHEMICALS INC Tirofiban 25MG
SDP
Catalog No. 502876042
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tirofiban (L700462) (MK-383) is a selective palate GPIIb/IIIa antagonist which inhibits platelet aggregation with IC50 of 9 nM. Purity 99.87%
Catalog No. 50-287-6042 Supplier TARGETMOL CHEMICALS INC Supplier No. T618225MG
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tirofiban (L700462) (MK-383) is a selective palate GPIIb/IIIa antagonist which inhibits platelet aggregation with IC50 of 9 nM. Purity 99.87%