Learn More
Apexbio Technology LLC Tirofiban, 25mg Cas# 144494-65-5
SDP
Supplier:  Apexbio Technology LLC B205225
Encompass_Preferred
Tirofiban 5mg. 1616113-45-1. MFCD28144726 Tirofiban (MK-383) is a selective palate GPIIb/IIIa antagonist which inhibits platelet aggregation with IC50 of 9 nM. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8678