Learn More
Selleck Chemical LLC Tirofiban Hydrochloride 10mg 150915-40-5 MK-383 Hydrochloride
SDP
Supplier:  Selleck Chemical LLC S859410MG
Encompass_Preferred
Tirofiban Hydrochloride (MK-383) is the hydrochloride salt form of tirofiban, a selective platelet GPIIb/IIIa antagonist which inhibits platelet aggregation. It is more soluable than Tirofiban. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8326