Learn More
TARGETMOL CHEMICALS INC TIROFIBAN HYDROCHLORIDE 25MG
SDP
Catalog No. 502577097
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tirofiban hydrochloride monohydrate (MK-383 Hydrochloride) is a potent non-peptide glycoprotein IIb/IIIa (integrins alphaIIbbetaIII) antagonist. purity: 99%
Catalog No. 50-257-7097 Supplier TARGETMOL CHEMICALS INC Supplier No. T253725MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tirofiban hydrochloride monohydrate (MK-383 Hydrochloride) is a potent non-peptide glycoprotein IIb/IIIa (integrins alphaIIbbetaIII) antagonist. purity: 99%