Learn More
Medchemexpress LLC Tirofiban (hydrochloride monohydrate) | 150915-40-5 | 99.9% | 495.07 | 5 MG
SDP
Catalog No. 5000364124
Encompass_Preferred
Tirofiban hydrochloride monohydrate is a selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist that inhibits fibrinogen binding to this receptor and has antithrombotic activity. It induces proliferation and migration on endothelial cells by inducing production of VEGF. Tirofiban hydrochloride monohydrate can significantly reduce myocardial no-reflow and ischemia-reperfusion injury by alleviating myocardial microvascular structural and endothelial dysfunction in the ischemic area.
  • Selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist
  • Inhibits fibrinogen binding to the receptor
  • Possesses antithrombotic activity
  • Induces proliferation and migration on endothelial cells
  • Significantly reduces myocardial no-reflow
  • Alleviates myocardial microvascular structural and endothelial dysfunction
Catalog No. 50-003-64124 Supplier Medchemexpress LLC Supplier No. 5MGHY17369
Tirofiban hydrochloride monohydrate is a selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist that inhibits fibrinogen binding to this receptor and has antithrombotic activity. It induces proliferation and migration on endothelial cells by inducing production of VEGF. Tirofiban hydrochloride monohydrate can significantly reduce myocardial no-reflow and ischemia-reperfusion injury by alleviating myocardial microvascular structural and endothelial dysfunction in the ischemic area.
  • Selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist
  • Inhibits fibrinogen binding to the receptor
  • Possesses antithrombotic activity
  • Induces proliferation and migration on endothelial cells
  • Significantly reduces myocardial no-reflow
  • Alleviates myocardial microvascular structural and endothelial dysfunction