Learn More
Medchemexpress LLC Tirofiban hydrochloride monohydrate | 150915-40-5 | 99.9% | 495.07 | 500 MG
SDP
Catalog No. 5000385996
Encompass_Preferred
Tirofiban hydrochloride monohydrate is a selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist. It inhibits fibrinogen binding to this receptor and demonstrates antithrombotic activity. This compound also induces proliferation and migration on endothelial cells by stimulating VEGF production, and can significantly reduce myocardial no-reflow and ischemia-reperfusion injury by improving myocardial microvascular structural and endothelial dysfunction in ischemic areas.
  • Selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist
  • Inhibits fibrinogen binding
  • Antithrombotic activity
  • Induces proliferation and migration on endothelial cells
  • Stimulates VEGF production
  • Reduces myocardial no-reflow and ischemia-reperfusion injury
  • Alleviates myocardial microvascular dysfunction
Catalog No. 50-003-85996 Supplier Medchemexpress LLC Supplier No. HY17369500MG
Tirofiban hydrochloride monohydrate is a selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist. It inhibits fibrinogen binding to this receptor and demonstrates antithrombotic activity. This compound also induces proliferation and migration on endothelial cells by stimulating VEGF production, and can significantly reduce myocardial no-reflow and ischemia-reperfusion injury by improving myocardial microvascular structural and endothelial dysfunction in ischemic areas.
  • Selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist
  • Inhibits fibrinogen binding
  • Antithrombotic activity
  • Induces proliferation and migration on endothelial cells
  • Stimulates VEGF production
  • Reduces myocardial no-reflow and ischemia-reperfusion injury
  • Alleviates myocardial microvascular dysfunction