Learn More
Medchemexpress LLC Tirofiban (hydrochloride monohydrate) | 150915-40-5 | MFCD07368623 | 99.9% | 495.07 g/mol | C22H39ClN2O6S | 10 MG
SDP
Catalog No. 5000298564
Encompass_Preferred
Tirofiban hydrochloride monohydrate is the hydrochloride monohydrate salt of tirofiban, a selective and reversible platelet integrin (Gp IIb/IIIa) antagonist supplied for research and analytical applications. It inhibits fibrinogen binding to the Gp IIb/IIIa receptor and is provided as a high-purity solid suitable for in vitro studies.
  • Selective, reversible Gp IIb/IIIa antagonist suitable for platelet research.
  • Inhibits fibrinogen binding to the Gp IIb/IIIa receptor.
  • High purity (≈99.9%) for reproducible experimental results.
  • Available in small laboratory quantities for analytical and preparative use.
  • Supplied as a stable hydrochloride monohydrate salt for consistent handling.
Catalog No. 50-002-98564 Supplier Medchemexpress LLC Supplier No. HY1736910MG
Tirofiban hydrochloride monohydrate is the hydrochloride monohydrate salt of tirofiban, a selective and reversible platelet integrin (Gp IIb/IIIa) antagonist supplied for research and analytical applications. It inhibits fibrinogen binding to the Gp IIb/IIIa receptor and is provided as a high-purity solid suitable for in vitro studies.
  • Selective, reversible Gp IIb/IIIa antagonist suitable for platelet research.
  • Inhibits fibrinogen binding to the Gp IIb/IIIa receptor.
  • High purity (≈99.9%) for reproducible experimental results.
  • Available in small laboratory quantities for analytical and preparative use.
  • Supplied as a stable hydrochloride monohydrate salt for consistent handling.