Learn More
Cayman Chemical TIrofIbnhydrochLRIde hydr 10mg
SDP
Catalog No. 502986466
Encompass_Preferred
A GPIIb/IIIa antagonist (IC50 = 5 nM in a radioligand binding assay); inhibits GPIIb/IIIa-dependent platelet aggregation (IC50 = 11 nM); inhibits HUVEC attachment to microtiter plates containing Fg, Vn, or Fn at concentrations >10,000-fold above the IC50 for platelet aggregation; prevents occlusive thrombosis and reduces thrombus mass in a canine model of electrically-induced coronary artery occlusive thrombosis
Catalog No. 50-298-6466 Supplier Cayman Chemical Supplier No. 2339210 MG
A GPIIb/IIIa antagonist (IC50 = 5 nM in a radioligand binding assay); inhibits GPIIb/IIIa-dependent platelet aggregation (IC50 = 11 nM); inhibits HUVEC attachment to microtiter plates containing Fg, Vn, or Fn at concentrations >10,000-fold above the IC50 for platelet aggregation; prevents occlusive thrombosis and reduces thrombus mass in a canine model of electrically-induced coronary artery occlusive thrombosis