Learn More
Abcam Tiron, supero x ide scavenger and antio x idant, 25G
Catalog No. 502704815
Encompass_Preferred
MW 314.2 Da. Cell permeable superoxide scavenger and antioxidant. Also used as indicator for complexometry. Can form complexes with titanium and iron.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4815 Supplier Abcam Supplier No. AB14623425G
MW 314.2 Da. Cell permeable superoxide scavenger and antioxidant. Also used as indicator for complexometry. Can form complexes with titanium and iron.
The product is subject to the following: Abcam Restricted Use Statement