Learn More
Selleck Chemical LLC Tirzepatide LY3298176-1mg
SDP
Catalog No. 502389875
Encompass_Preferred
Tirzepatide (LY3298176, GIP/GLP-1 RA, TZP) is a dual GIP/GLP-1 receptor agonist. Tirzepatide differentially induces internalization of the GIP and GLP-1 receptors with EC50 values of 18.2 nM and 18.1 nM, respectively.
Catalog No. 50-238-9875 Supplier Selleck Chemical LLC Supplier No. P12061MG
Tirzepatide (LY3298176, GIP/GLP-1 RA, TZP) is a dual GIP/GLP-1 receptor agonist. Tirzepatide differentially induces internalization of the GIP and GLP-1 receptors with EC50 values of 18.2 nM and 18.1 nM, respectively.