Learn More
Cayman Chemical TIrzepatIdesodIum salt 10mg
SDP
Catalog No. 502942003
Encompass_Preferred
A dual GLP-1R and GIP receptor agonist; induces cAMP production in HEK293 cells expressing human GLP-1R or human GIP receptor (EC50s = 6.54 and 1.01 nM, respectively); induces receptor internalization in HEK293 cells expressing human GLP-1R or GIP receptor at 100 nM; decreases body weight, food intake, plasma levels of leptin, triglycerides, and FFAs, hepatic levels of triglycerides, and blood glucose levels in a mouse model of high-fat diet-induced obesity at 10 nmol/kg per day; prevents A. alternata-induced increases in the numbers of eosinophils and lymphocytes levels in the BALF from A. alternata-challenged mice at 50 nmol/kg every three days; inhibits methacholine-induced bronchoconstriction in a mouse model of diabetes-induced asthma at 50 nmol/kg every three days
Catalog No. 50-294-2003 Supplier Cayman Chemical Supplier No. 3974810 MG
A dual GLP-1R and GIP receptor agonist; induces cAMP production in HEK293 cells expressing human GLP-1R or human GIP receptor (EC50s = 6.54 and 1.01 nM, respectively); induces receptor internalization in HEK293 cells expressing human GLP-1R or GIP receptor at 100 nM; decreases body weight, food intake, plasma levels of leptin, triglycerides, and FFAs, hepatic levels of triglycerides, and blood glucose levels in a mouse model of high-fat diet-induced obesity at 10 nmol/kg per day; prevents A. alternata-induced increases in the numbers of eosinophils and lymphocytes levels in the BALF from A. alternata-challenged mice at 50 nmol/kg every three days; inhibits methacholine-induced bronchoconstriction in a mouse model of diabetes-induced asthma at 50 nmol/kg every three days