Learn More
Andwin Scientific TIS-TEK MOLD RELEAS AERO 12/CS
SDP
Catalog No. NC9765672
Encompass
The Tissue-Tek Mold Release Concentrate is ideal for coating base molds to ensure clean separation of embedding paraffin from mold. This product is packaged in a 4 oz. bottle. It is sold in a case of 12.
Catalog No. NC9765672 Supplier Andwin Scientific Supplier No. 4141
The Tissue-Tek Mold Release Concentrate is ideal for coating base molds to ensure clean separation of embedding paraffin from mold. This product is packaged in a 4 oz. bottle. It is sold in a case of 12.