Learn More
Aqua Solutions TISAB Buffer, Special
SDP
Catalog No. NC1746668
Encompass_Preferred
Compositon: 5.58% Acetic Acid (64-19-7), 11.701% Sodium Chloride (7647-14-5), 1.116% Sodium Citrate Dihydrate (6132-04-3), 81.604% Water (7732-18-5)
Catalog No. NC1746668 Supplier Aqua Solutions Supplier No. HON136
Compositon: 5.58% Acetic Acid (64-19-7), 11.701% Sodium Chloride (7647-14-5), 1.116% Sodium Citrate Dihydrate (6132-04-3), 81.604% Water (7732-18-5)