Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TISAB Concentrate, Ricca Chemical
SDP

Catalog No. 01213123 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-213-123 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-213-123 Supplier Ricca Chemical Company Supplier No. R8670500500A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material TISAB Concentrate
Packaging 500 mL HDPE Bottle
CAS 7732-18-5, 64-19-7, 1310-73-2, 13291-61-7, 7732-18-5
Certificate of Analysis Yes
CAS Min % 0.49%, 73.58%, 7.93%, 18.01%
CAS Max % 7.93%, 73.58%, 18.01%, 0.49%
Traceability to NIST NIST SRM# 185i, 186-I-g, 186-II-g
Physical Form Liquid
Color Colorless
pH 5.30 to 5.50
Melting Point Approximately 0°C
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Solution
Type Standard
Quantity 500 mL
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.