Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TISAB II Solution , MilliporeSigma™ Supelco™

Catalog No. 111002582 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2582 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2582 Supplier MilliporeSigma Supelco Supplier No. 89466500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TISAB, in general, is mixed with samples, as it helps in maintaining the ionic strength and pH constant. It is mainly used to decomplex metal-F complexes. TISAB II can be prepared using NaCl with glacial acetic acid, 1,2-Cyclohexylenedinitrilotetraacetic acid and NaOH. It also has high concentrations of cation chelators that hydrolyse fluoride-ion pairs to form fluoride ions.

TRUSTED_SUSTAINABILITY

Specifications

Density 1.18 g/mL (at 20°C)
Synonym Total ionic strength adjustment buffer II solution
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.