Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TISAB III Solution with CDTA, Reagents

Catalog No. 01333431
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-431 1 L
01-333-430 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-333-431 Supplier Reagents Holdings Llc Supplier No. CT1315001A
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TISAB III Solution with CDTA, Reagent, use for Water analysis (Fluoride). Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material TISAB III Solution with CDTA
CAS 7732-18-5, 12125-02-9, 631-61-8
Boiling Point 100°C
Color Colorless
For Use With (Equipment) Fluoride Meter
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.1269
Sterility Non-sterile
Synonym Total Ionic Strength Adjustment Buffer
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.